Learn More
Medchemexpress LLC 1-O-Hexadecyl-sn-glycerol | 506-03-6 | 99.9% | 316.53 | 25 MG
SDP
Catalog No. 5000366677
Encompass_Preferred
1-O-Hexadecyl-sn-glycerol [(S)-HXDG] is an alkyl glycerol monoether derived from *T. aurantia*. It is for research use only.
  • Alkyl glycerol monoether
  • Derived from *T. aurantia*
  • Powder can be stored at -20°C for up to 3 years or at 4°C for up to 2 years
  • In solvent, can be stored at -80°C for up to 6 months or at -20°C for up to 1 month
  • Solubility in DMSO: 25 mg/mL (78.98 mM), requires ultrasonic assistance
  • For in vivo use, can be prepared by adding 10% DMSO followed by 90% corn oil, resulting in a clear solution with solubility ≥ 2.5 mg/mL (7.90 mM)
Catalog No. 50-003-66677 Supplier Medchemexpress LLC Supplier No. 25MGHYW011309A
1-O-Hexadecyl-sn-glycerol [(S)-HXDG] is an alkyl glycerol monoether derived from *T. aurantia*. It is for research use only.
  • Alkyl glycerol monoether
  • Derived from *T. aurantia*
  • Powder can be stored at -20°C for up to 3 years or at 4°C for up to 2 years
  • In solvent, can be stored at -80°C for up to 6 months or at -20°C for up to 1 month
  • Solubility in DMSO: 25 mg/mL (78.98 mM), requires ultrasonic assistance
  • For in vivo use, can be prepared by adding 10% DMSO followed by 90% corn oil, resulting in a clear solution with solubility ≥ 2.5 mg/mL (7.90 mM)