Learn More
Medchemexpress LLC 1-O-Methyl-2-2-deoxy-D-ribose | 60134-26-1 | C6H12O4 | 25 G
SDP
Catalog No. 5000357557
Encompass_Preferred
1-O-Methyl-2-deoxy-D-ribose is a ribose derivative obtained through a one-step reaction by introducing a methoxy protective group at the anomeric carbon position under acidic conditions, which assists in the acquisition of 2-deoxy-D-ribose. It can be used in research for the synthesis of chemical materials.
  • Ribose derivative
  • One-step synthesis
  • Facilitates 2-deoxy-D-ribose acquisition
  • Research utility in chemical materials synthesis
  • Available documentation: Data sheet, SDS, and handling instructions
Catalog No. 50-003-57557 Supplier Medchemexpress LLC Supplier No. 25GHY77957
1-O-Methyl-2-deoxy-D-ribose is a ribose derivative obtained through a one-step reaction by introducing a methoxy protective group at the anomeric carbon position under acidic conditions, which assists in the acquisition of 2-deoxy-D-ribose. It can be used in research for the synthesis of chemical materials.
  • Ribose derivative
  • One-step synthesis
  • Facilitates 2-deoxy-D-ribose acquisition
  • Research utility in chemical materials synthesis
  • Available documentation: Data sheet, SDS, and handling instructions