Learn More
Medchemexpress LLC 1-O-Methyl-2-deoxy-D-ribose | 60134-26-1 | C6H12O4 | 10 G
SDP
Catalog No. 5000432739
Encompass_Preferred
1-O-Methyl-2-deoxy-D-ribose is a ribose derivative obtained via a one-step reaction by introducing a methoxy protective group at the anomeric carbon position under acidic conditions, facilitating the acquisition of 2-deoxy-D-ribose. It is utilized in research on the synthesis of chemical materials.
  • Ribose derivative
  • Obtained through a one-step reaction
  • Facilitates acquisition of 2-deoxy-D-ribose
  • Utilized in research on chemical material synthesis
Catalog No. 50-004-32739 Supplier Medchemexpress LLC Supplier No. HY7795710G
1-O-Methyl-2-deoxy-D-ribose is a ribose derivative obtained via a one-step reaction by introducing a methoxy protective group at the anomeric carbon position under acidic conditions, facilitating the acquisition of 2-deoxy-D-ribose. It is utilized in research on the synthesis of chemical materials.
  • Ribose derivative
  • Obtained through a one-step reaction
  • Facilitates acquisition of 2-deoxy-D-ribose
  • Utilized in research on chemical material synthesis