Learn More
Medchemexpress LLC 1-O-Methyl-2-deoxy-D-ribose | 60134-26-1 | C6H12O4 | 100 G
SDP
Catalog No. 5000433465
Encompass_Preferred
1-O-Methyl-2-deoxy-D-ribose is a ribose derivative formed by introducing a methoxy protective group at the anomeric carbon position under acidic conditions. This process facilitates the acquisition of 2-deoxy-D-ribose and can be utilized in research on the synthesis of chemical materials.
  • Ribose derivative obtained through a one-step reaction
  • Methoxy protective group introduced at anomeric carbon position
  • Facilitates the acquisition of 2-deoxy-D-ribose
  • Utilized in research on the synthesis of chemical materials
  • For research use only
Catalog No. 50-004-33465 Supplier Medchemexpress LLC Supplier No. HY77957100G
1-O-Methyl-2-deoxy-D-ribose is a ribose derivative formed by introducing a methoxy protective group at the anomeric carbon position under acidic conditions. This process facilitates the acquisition of 2-deoxy-D-ribose and can be utilized in research on the synthesis of chemical materials.
  • Ribose derivative obtained through a one-step reaction
  • Methoxy protective group introduced at anomeric carbon position
  • Facilitates the acquisition of 2-deoxy-D-ribose
  • Utilized in research on the synthesis of chemical materials
  • For research use only