Learn More
Medchemexpress LLC 1-O-Methyl-2-deoxy-D-ribose | 60134-26-1 | C6H12O4 | 5 G
SDP
Catalog No. 5000434946
Encompass_Preferred
1-O-Methyl-2-deoxy-D-ribose is a ribose derivative that can be obtained through a one-step reaction by introducing a methoxy protective group at the anomeric carbon position under acidic conditions. This facilitates the acquisition of 2-deoxy-D-ribose and can be utilized in research on the synthesis of chemical materials.
  • Ribose derivative
  • Obtained via one-step reaction
  • Features a methoxy protective group at the anomeric carbon
  • Facilitates the acquisition of 2-deoxy-D-ribose
  • Suitable for research on the synthesis of chemical materials
Catalog No. 50-004-34946 Supplier Medchemexpress LLC Supplier No. HY779575G
1-O-Methyl-2-deoxy-D-ribose is a ribose derivative that can be obtained through a one-step reaction by introducing a methoxy protective group at the anomeric carbon position under acidic conditions. This facilitates the acquisition of 2-deoxy-D-ribose and can be utilized in research on the synthesis of chemical materials.
  • Ribose derivative
  • Obtained via one-step reaction
  • Features a methoxy protective group at the anomeric carbon
  • Facilitates the acquisition of 2-deoxy-D-ribose
  • Suitable for research on the synthesis of chemical materials