Learn More
Medchemexpress LLC 1-O-Methyl-2-deoxy-D-ribose | 60134-26-1 | C6H12O4 | 50 G
SDP
Catalog No. 5000433395
Encompass_Preferred
1-O-Methyl-2-deoxy-D-ribose is a ribose derivative that can be conveniently obtained through a one-step reaction by introducing a methoxy protective group at the anomeric carbon position under acidic conditions. This facilitates the acquisition of 2-deoxy-D-ribose.
  • Utilized in research on the synthesis of chemical materials.
Catalog No. 50-004-33395 Supplier Medchemexpress LLC Supplier No. HY7795750G
1-O-Methyl-2-deoxy-D-ribose is a ribose derivative that can be conveniently obtained through a one-step reaction by introducing a methoxy protective group at the anomeric carbon position under acidic conditions. This facilitates the acquisition of 2-deoxy-D-ribose.
  • Utilized in research on the synthesis of chemical materials.