Learn More
Alkali Scientific 1 O Methyl α D mannopyranoside [Methyl α D Mannopyranoside], 100 G
SDP
Catalog No. 5000659627
Encompass_Preferred
Methyl α-D-Mannopyranoside (100g) is a high-quality carbohydrate reagent used in laboratory experiments that require a stable and reliable source of this sugar derivative. Methyl α-D-Mannopyranoside is used in biochemical research for the study of glycosidic bonds, carbohydrate-protein interactions, and in the synthesis of oligosaccharides and glycoproteins. This reagent is commonly employed in research on carbohydrate chemistry, enzymology, and cell biology. The 100g bottle is ideal for medium to large-scale experiments, offering an essential carbohydrate derivative for researchers studying molecular interactions, glycosylation, and other sugar-related biochemical processes.
Catalog No. 50-006-59627 Supplier Alkali Scientific Supplier No. HYL001CP
Methyl α-D-Mannopyranoside (100g) is a high-quality carbohydrate reagent used in laboratory experiments that require a stable and reliable source of this sugar derivative. Methyl α-D-Mannopyranoside is used in biochemical research for the study of glycosidic bonds, carbohydrate-protein interactions, and in the synthesis of oligosaccharides and glycoproteins. This reagent is commonly employed in research on carbohydrate chemistry, enzymology, and cell biology. The 100g bottle is ideal for medium to large-scale experiments, offering an essential carbohydrate derivative for researchers studying molecular interactions, glycosylation, and other sugar-related biochemical processes.