Learn More
Alkali Scientific 1 O Methyl α D mannopyranoside [Methyl α D Mannopyranoside], 500 G
SDP
Catalog No. 5000659580
Encompass_Preferred
Methyl α-D-mannopyranoside (HYL003) is a reagent commonly used in biochemical and laboratory applications. This compound is a methylated form of D-mannose, a naturally occurring sugar. The α-anomer is preferred in many experiments for its stability and reactivity. Methyl α-D-mannopyranoside is widely used as a glycosylation agent in the synthesis of complex oligosaccharides and glycoproteins. It also serves as a reference compound for determining the structure of carbohydrates. The 500g bottle size is ideal for laboratory use where large quantities are needed for various experiments in carbohydrate chemistry, biochemistry, and structural biology.
Catalog No. 50-006-59580 Supplier Alkali Scientific Supplier No. HYL003CP
Add to Cart
Add to Cart
Methyl α-D-mannopyranoside (HYL003) is a reagent commonly used in biochemical and laboratory applications. This compound is a methylated form of D-mannose, a naturally occurring sugar. The α-anomer is preferred in many experiments for its stability and reactivity. Methyl α-D-mannopyranoside is widely used as a glycosylation agent in the synthesis of complex oligosaccharides and glycoproteins. It also serves as a reference compound for determining the structure of carbohydrates. The 500g bottle size is ideal for laboratory use where large quantities are needed for various experiments in carbohydrate chemistry, biochemistry, and structural biology.