Learn More
Medchemexpress LLC 1-O-n-Octyl-B-D-gluc 1mg | 129522-81-2 | 309.47 g·mol^-1 | C14H11D17O6 | 1 MG
SDP
Catalog No. 5000229222
Encompass_Preferred
Deuterated n-octyl β-D-glucopyranoside (d17) is a deuterium-labeled non-ionic detergent used to solubilize membrane proteins and as an analytical standard in quantitative methods. It is supplied in small, research-scale quantities for biochemical and analytical workflows.
  • Deuterium-labeled non-ionic detergent for membrane protein solubilization.
  • Serves as a tracer and internal standard for quantitative NMR, GC-MS, and LC-MS.
  • Supplied in small, research-grade quantities suitable for analytical use.
  • Deuteration provides a mass shift for clear distinction from unlabeled analogs in mass spectrometry.
  • Batch-specific certificate of analysis available for purity and isotopic enrichment verification.
Catalog No. 50-002-29222 Supplier Medchemexpress LLC Supplier No. HY116285S11MG
Deuterated n-octyl β-D-glucopyranoside (d17) is a deuterium-labeled non-ionic detergent used to solubilize membrane proteins and as an analytical standard in quantitative methods. It is supplied in small, research-scale quantities for biochemical and analytical workflows.
  • Deuterium-labeled non-ionic detergent for membrane protein solubilization.
  • Serves as a tracer and internal standard for quantitative NMR, GC-MS, and LC-MS.
  • Supplied in small, research-grade quantities suitable for analytical use.
  • Deuteration provides a mass shift for clear distinction from unlabeled analogs in mass spectrometry.
  • Batch-specific certificate of analysis available for purity and isotopic enrichment verification.