Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-O-n-Octyl-B-D-gluc 5mg | 129522-81-2 | 5 MG
SDP

Catalog No. 5000227338
Encompass_Preferred

1-O-n-Octyl-β-D-glucopyranoside-d17 (CAS 129522-81-2) is a deuterium-labeled n-octyl β-D-glucopyranoside used as a non-ionic surfactant and analytical tracer in biochemical and biophysical research. Supplied as a solid in small mass packaging for analytical and tracer studies.

  • Deuterium-labeled n-octyl β-D-glucopyranoside for tracer applications.
  • Non-ionic surfactant useful for membrane protein solubilization.
  • Provided as a solid in small-mass packaging suitable for analytical workflows.
  • Molecular weight 309.47 g·mol⁻¹ and formula C14H11D17O6.
  • Documentation available: data sheet and safety data sheet on request.

Catalog No. 50-002-27338 Supplier Medchemexpress LLC Supplier No. HY116285S15MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-O-n-Octyl-β-D-glucopyranoside-d17 (CAS 129522-81-2) is a deuterium-labeled n-octyl β-D-glucopyranoside used as a non-ionic surfactant and analytical tracer in biochemical and biophysical research. Supplied as a solid in small mass packaging for analytical and tracer studies.

  • Deuterium-labeled n-octyl β-D-glucopyranoside for tracer applications.
  • Non-ionic surfactant useful for membrane protein solubilization.
  • Provided as a solid in small-mass packaging suitable for analytical workflows.
  • Molecular weight 309.47 g·mol⁻¹ and formula C14H11D17O6.
  • Documentation available: data sheet and safety data sheet on request.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.