Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical 1-(o-Ph-NO2)-3-(DMTr)oxypropyl amidite | 200 mg | CAS 168329-40-6 | InChIKey KHOFWXQDBRVNMM-UHFFFAOYSA-N

Catalog No. 502363801
Encompass

1-(o-Ph-NO2)-3-(DMTr)oxypropyl amidite is a Amidite reagent (Subcategory: Spacer Modifiers) sold by WuXi TIDES. Offered in 200 mg. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 168329-40-6
- MDL: No data
- InChIKey: KHOFWXQDBRVNMM-UHFFFAOYSA-N
- Molecular Weight: 699.784761998
- Molecular Formula: C39H46N3O7P
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 200 mg
- Net Weight: 0.2 g
- Gross Weight: 6.9 g
- Commodity Code: 29299090
- Country Of Origin: China
- IUPAC: 3-(bis(4-methoxyphenyl)(phenyl)methoxy)-1-(2-nitrophenyl)propyl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(OC(C1=C(C=CC=C1)[N+]([O-])=O)CCOC(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)C4=CC=CC=C4)OCCC#N)C(C)C)C

Catalog No. 50-236-3801 Supplier STA Pharmaceutical Supplier No. STAAM10654200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-(o-Ph-NO2)-3-(DMTr)oxypropyl amidite is a Amidite reagent (Subcategory: Spacer Modifiers) sold by WuXi TIDES. Offered in 200 mg. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 168329-40-6
- MDL: No data
- InChIKey: KHOFWXQDBRVNMM-UHFFFAOYSA-N
- Molecular Weight: 699.784761998
- Molecular Formula: C39H46N3O7P
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 200 mg
- Net Weight: 0.2 g
- Gross Weight: 6.9 g
- Commodity Code: 29299090
- Country Of Origin: China
- IUPAC: 3-(bis(4-methoxyphenyl)(phenyl)methoxy)-1-(2-nitrophenyl)propyl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(OC(C1=C(C=CC=C1)[N+]([O-])=O)CCOC(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)C4=CC=CC=C4)OCCC#N)C(C)C)C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.