Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-O-trans-Cinnamoyl-β-D-glucopyranose | 40004-96-4 | 310.30 | 1 MG
SDP

Catalog No. 5000415077
Encompass_Preferred

1-O-trans-Cinnamoyl-β-D-glucopyranose is a natural product isolated from fruits, intended for research use only. This compound is characterized by its high purity and specific chemical structure, making it suitable for various laboratory applications.

  • High purity of 99.65%
  • Molecular weight of 310.30
  • Chemical formula C15H18O7
  • CAS number 40004-96-4
  • Appears as a white to off-white solid
  • Classified as a phenylpropanoid
  • Derived from plants, specifically Rosaceae and Spiraea thunbergii Siebold ex Blume
  • Stable for 3 years as a powder at -20°C
  • Stable for 6 months in solvent at -80°C

Catalog No. 50-004-15077 Supplier Medchemexpress LLC Supplier No. HYN116311MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-O-trans-Cinnamoyl-β-D-glucopyranose is a natural product isolated from fruits, intended for research use only. This compound is characterized by its high purity and specific chemical structure, making it suitable for various laboratory applications.

  • High purity of 99.65%
  • Molecular weight of 310.30
  • Chemical formula C15H18O7
  • CAS number 40004-96-4
  • Appears as a white to off-white solid
  • Classified as a phenylpropanoid
  • Derived from plants, specifically Rosaceae and Spiraea thunbergii Siebold ex Blume
  • Stable for 3 years as a powder at -20°C
  • Stable for 6 months in solvent at -80°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.