Learn More
Medchemexpress LLC 1-Octacosanol | 557-61-9 | 250 MG
SDP
Catalog No. 5000420709
Encompass_Preferred
1-Octacosanol is a straight-chain aliphatic 28-carbon fatty alcohol and a major component of Policosanol. It is known for its diverse biological activities. This product appears as a white to off-white solid with a purity of 98.0%. It has a molecular weight of 410.77 and a chemical formula of C28H58O. It is insoluble or slightly soluble in DMSO, insoluble in Ethanol, and insoluble in H2O, and is usually formulated as a suspension.
  • Exhibits anti-fatigue effects
  • Demonstrates anti-angiogenic properties
  • Has cholesterol-lowering effects
  • Shows insecticidal activity against *Spodoptera frugiperda*
Catalog No. 50-004-20709 Supplier Medchemexpress LLC Supplier No. 250MGHYN6811
1-Octacosanol is a straight-chain aliphatic 28-carbon fatty alcohol and a major component of Policosanol. It is known for its diverse biological activities. This product appears as a white to off-white solid with a purity of 98.0%. It has a molecular weight of 410.77 and a chemical formula of C28H58O. It is insoluble or slightly soluble in DMSO, insoluble in Ethanol, and insoluble in H2O, and is usually formulated as a suspension.
  • Exhibits anti-fatigue effects
  • Demonstrates anti-angiogenic properties
  • Has cholesterol-lowering effects
  • Shows insecticidal activity against *Spodoptera frugiperda*