Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-octadecanamine, n,n-dimethyl- | 124-28-7 | 97.0% | 250 MG
SDP

Catalog No. 5000232932
Encompass_Preferred

Dymanthine (N,N-dimethyloctadecan-1-amine) is an aliphatic tertiary amine supplied as a research reagent for studies of intestinal helminth infestations. It is characterized by a long alkyl chain and a tertiary amine functional group, and is provided at high reported purity for analytical and biological research.

  • Chemical name: 1-octadecanamine, n,n-dimethyl-.
  • Cas number: 124-28-7.
  • Molecular formula: C20H43N.
  • Molecular weight: 297.56.
  • Purity: 97.0% by NMR.
  • Physical state: Solid below 23°C, liquid above 23°C.
  • Intended use: Research reagent for studies of intestinal helminth infestations.
  • Packaging: available in multiple pack sizes.

Catalog No. 50-002-32932 Supplier Medchemexpress LLC Supplier No. HY121429250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Dymanthine (N,N-dimethyloctadecan-1-amine) is an aliphatic tertiary amine supplied as a research reagent for studies of intestinal helminth infestations. It is characterized by a long alkyl chain and a tertiary amine functional group, and is provided at high reported purity for analytical and biological research.

  • Chemical name: 1-octadecanamine, n,n-dimethyl-.
  • Cas number: 124-28-7.
  • Molecular formula: C20H43N.
  • Molecular weight: 297.56.
  • Purity: 97.0% by NMR.
  • Physical state: Solid below 23°C, liquid above 23°C.
  • Intended use: Research reagent for studies of intestinal helminth infestations.
  • Packaging: available in multiple pack sizes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.