Learn More
Medchemexpress LLC 1-Octadecanol | 112-92-5 | MFCD00002823 | 1000g
SDP
Catalog No. 5000266236
Encompass_Preferred
1-Hydroxyoctadecane (Stearyl alcohol) is a long-chain fatty alcohol and excipient 1-Hydroxyoctadecane forms a closed oil film thereby reducing transepidermal water loss (TEWL) and promoting skin moisturization 1-Hydroxyoctadecane improves Imiquimod (HY-B0180)-induced psoriatic skin inflammation in mice[1]
Catalog No. 50-002-66236 Supplier Medchemexpress LLC Supplier No. HYY18091000G
1-Hydroxyoctadecane (Stearyl alcohol) is a long-chain fatty alcohol and excipient 1-Hydroxyoctadecane forms a closed oil film thereby reducing transepidermal water loss (TEWL) and promoting skin moisturization 1-Hydroxyoctadecane improves Imiquimod (HY-B0180)-induced psoriatic skin inflammation in mice[1]