Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Octadecyl (2Z)-2-butenedioate | 2424-62-6 | MFCD08274815 | ≥98.0% | 368.55 g/mol | C22H40O4 | 500 MG
SDP

Catalog No. 5000282701
Encompass_Preferred

Monostearyl maleate (1-Octadecyl (2Z)-2-butenedioate) is a long-chain alkyl ester used as a specialty reagent and intermediate in chemical synthesis and formulation research. It is supplied as a white to off-white solid and characterized for analytical and preparative work.

  • High purity: ≥98.0%.
  • Molecular formula: C22H40O4.
  • Molecular weight: 368.55 g/mol.
  • Appearance: white to off-white solid.
  • Storage: store at room temperature; in solvent, -80°C (2 years) or -20°C (1 year).
  • Analytical data: 1H NMR consistent with structure.
  • Typical uses: ester intermediate and formulation reagent for research applications.

Catalog No. 50-002-82701 Supplier Medchemexpress LLC Supplier No. HYW140480500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monostearyl maleate (1-Octadecyl (2Z)-2-butenedioate) is a long-chain alkyl ester used as a specialty reagent and intermediate in chemical synthesis and formulation research. It is supplied as a white to off-white solid and characterized for analytical and preparative work.

  • High purity: ≥98.0%.
  • Molecular formula: C22H40O4.
  • Molecular weight: 368.55 g/mol.
  • Appearance: white to off-white solid.
  • Storage: store at room temperature; in solvent, -80°C (2 years) or -20°C (1 year).
  • Analytical data: 1H NMR consistent with structure.
  • Typical uses: ester intermediate and formulation reagent for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.