Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Octadecyl-4-(4-phenyl-1,3-butadienyl)pyridinium Bromide 96.0+%, TCI America™
SDP

Catalog No. O01931G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O01931G Supplier TCI America Supplier No. O01931G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 165678-32-0
Molecular Formula C33H50BrN
Molecular Weight (g/mol) 540.674
MDL Number MFCD00060045
InChI Key GERCFVKULBVBIO-UHFFFAOYSA-M
PubChem CID 53411749
IUPAC Name 1-octadecyl-4-(4-phenylbuta-1,3-dienyl)pyridin-1-ium;bromide
SMILES CCCCCCCCCCCCCCCCCC[N+]1=CC=C(C=C1)C=CC=CC2=CC=CC=C2.[Br-]

Specifications

Color Yellow
Quantity 1 g
Formula Weight 540.67
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Octadecyl-4-(4-phenyl-1,3-butadienyl)pyridinium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.