Learn More
Medchemexpress LLC 1-octanamine | 111-86-4 | MFCD00008247 | 99.7% | 129.24 g/mol | C8H19N | 500 G
SDP
Catalog No. 5000301602
Encompass_Preferred
Octylamine (1-octanamine) is a primary aliphatic amine reagent used as a building block in organic synthesis and biochemical research. It is a colorless to light yellow liquid (molecular formula C8H19N; molecular weight 129.24 g/mol) commonly used as an intermediate for surfactants, corrosion inhibitors, and other amine-derived products. Purity and storage instructions are lot-specific and provided on the certificate of analysis.
  • Primary aliphatic amine suitable for organic synthesis and derivatization.
  • Colorless to light yellow liquid, easy to dispense and handle.
  • Molecular formula C8H19N and molecular weight 129.24 g/mol.
  • High reported purity (about 99.7% GC); check COA for lot-specific value.
  • Commonly used as an intermediate for surfactants, corrosion inhibitors, and specialty chemicals.
  • Supplied with batch-specific certificate of analysis detailing purity and storage recommendations.
Catalog No. 50-003-01602 Supplier Medchemexpress LLC Supplier No. HYW032014500G
Octylamine (1-octanamine) is a primary aliphatic amine reagent used as a building block in organic synthesis and biochemical research. It is a colorless to light yellow liquid (molecular formula C8H19N; molecular weight 129.24 g/mol) commonly used as an intermediate for surfactants, corrosion inhibitors, and other amine-derived products. Purity and storage instructions are lot-specific and provided on the certificate of analysis.
  • Primary aliphatic amine suitable for organic synthesis and derivatization.
  • Colorless to light yellow liquid, easy to dispense and handle.
  • Molecular formula C8H19N and molecular weight 129.24 g/mol.
  • High reported purity (about 99.7% GC); check COA for lot-specific value.
  • Commonly used as an intermediate for surfactants, corrosion inhibitors, and specialty chemicals.
  • Supplied with batch-specific certificate of analysis detailing purity and storage recommendations.