Learn More
Medchemexpress LLC 1-Octanamine, N-octyl- | 1120-48-5 | 99.36% | 241.46 | 10 G
SDP
Catalog No. 5000403439
Encompass_Preferred
Dioctylamine (Di-n-octylamine) is a chemical reagent that features a secondary amine linked to two saturated C8 tails. This secondary amine acts as a versatile substrate, capable of building a wide variety of bonds. Compounds like Dioctylamine may be utilized as intermediates in the construction of lipids for application in lipid nanoparticles.
  • It is a chemical reagent featuring a secondary amine linked to two saturated C8 tails.
  • The secondary amine is a versatile substrate which can be used to build a wide variety of bonds.
  • It may be used as an intermediate in building lipids for use in lipid nanoparticles.
Catalog No. 50-004-03439 Supplier Medchemexpress LLC Supplier No. HY3441910G
Dioctylamine (Di-n-octylamine) is a chemical reagent that features a secondary amine linked to two saturated C8 tails. This secondary amine acts as a versatile substrate, capable of building a wide variety of bonds. Compounds like Dioctylamine may be utilized as intermediates in the construction of lipids for application in lipid nanoparticles.
  • It is a chemical reagent featuring a secondary amine linked to two saturated C8 tails.
  • The secondary amine is a versatile substrate which can be used to build a wide variety of bonds.
  • It may be used as an intermediate in building lipids for use in lipid nanoparticles.