Learn More
Medchemexpress LLC 1-Octanaminium, N,N,N-trioctyl-, bromide (1:1) | 14866-33-2 | MFCD00011863 | 98.0% | 546.79 g/mol | C32H68BrN | 500 G
SDP
Supplier:  Medchemexpress LLC HYW018574500G
Encompass_Preferred
Tetraoctylammonium bromide is a quaternary ammonium salt commonly used as a phase-transfer catalyst and reagent in organic synthesis. It is an off-white solid (C32H68BrN) with a molecular weight of about 546.79 g/mol, typically supplied at ≥98% purity for laboratory and synthetic applications.
  • Phase-transfer catalyst for enhancing reactions between immiscible phases.
  • Facilitates nucleophilic substitutions and anion transfer in organic synthesis.
  • High purity suitable for research and synthesis.
  • Solid form is easy to weigh and handle.
  • Compatible with common organic solvents and reagents.
Catalog No. 50-003-01871