Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Octanesulfonic acid, sodium salt (1:1) | 5324-84-5 | 97.0% | 216.27 | 25 G
SDP

Catalog No. 5000451363
Encompass_Preferred

Sodium octane-1-sulfonate is an ion-pairing agent that can be used as a buffer in biochemical research and HPLC. It is for research use only.

  • High purity: 99.92%
  • Application: Used as an ion-pairing agent
  • Functionality: Acts as a buffer
  • Research utility: Suitable for biochemical research and HPLC applications

Catalog No. 50-004-51363 Supplier Medchemexpress LLC Supplier No. 25GHYW008600
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium octane-1-sulfonate is an ion-pairing agent that can be used as a buffer in biochemical research and HPLC. It is for research use only.

  • High purity: 99.92%
  • Application: Used as an ion-pairing agent
  • Functionality: Acts as a buffer
  • Research utility: Suitable for biochemical research and HPLC applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.