Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Octanesulfonyl chloride, 97%, Thermo Scientific™

Catalog No. AAA1544018
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1544018 50 g
AAA1544030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1544018 Supplier Thermo Scientific Chemicals Supplier No. A1544018

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7795-95-1
Molecular Formula C8H17ClO2S
Molecular Weight (g/mol) 212.732
MDL Number MFCD00014729
InChI Key WIVNTNLDTMNDNO-UHFFFAOYSA-N
Synonym 1-octanesulfonyl chloride, n-octylsulfonyl chloride, octanesulfonyl chloride, unii-3sy5pr1ee7, 3sy5pr1ee7, 1-octanesulfonylchloride, 1-octanesulphonyl chloride, chlorooctylsulfone, octane sulfonyl chloride, acmc-209pcx
PubChem CID 82261
IUPAC Name octane-1-sulfonyl chloride
SMILES CCCCCCCCS(=O)(=O)Cl

Specifications

Melting Point 14°C
Density 1.089
Boiling Point 75°C (0.03 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.46
Linear Formula CH3(CH2)7SO2Cl
Quantity 50 g
UN Number UN3265
Beilstein 1763921
Sensitivity Moisture sensitive
Formula Weight 212.74
Percent Purity 97%
Chemical Name or Material 1-Octanesulfonyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 232-249-8
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.