Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Octanethiol

Catalog No. 4718362L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-183-62L 2 L
47-183-625ML 25 mL
47-183-6250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 47-183-62L Supplier Sigma Aldrich Supplier No. 4718362L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Octanethiol (OT) is C8 alkanethiol that forms self-assembled monolayer (SAM) on a variety of nanoparticles. It has a low flash point and low vapor pressure.OT can be used as a sulfur precursor for the preparation of shell for CdSe/CdS quantum dots. It forms ligands that help in enhancing the power efficiency of hetero-junction nanorod sensitized solar cells as it enhances the electron extraction from the nanorod and TiO2 interface. OT based stabilized coating on copper nanoparticles (CuNPs) may be used in the dispersion of these colloidal particles in different sample solutions. It can be used to improve charge injection by creating a monolayer on gold source-drain electrode surfaces for bottom-contact thin-film transistors (TFTs).Product of Arkema Inc.

Specifications

CAS 111-88-6
Density 0.843 g/mL (at 25°C)
Boiling Point 197°C to 200°C
Molecular Formula C8H18S
Refractive Index n20/D 1.452 (literature)
Linear Formula CH3(CH2)7 SH
Quantity 2 L
MDL Number MFCD00004912
Synonym n-Octyl mercaptan; Mercaptan C8; Octyl mercaptan
Molecular Weight (g/mol) 146.29
Percent Purity ≥98.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.