Learn More
Medchemexpress LLC 1-Octanol | 111-87-5 | MFCD00002988 | 98.0% | 130.23 g/mol | C8H18O | 1 G
SDP
Catalog No. 5000299357
Encompass_Preferred
1-Octanol (octan-1-ol) is a straight-chain primary fatty alcohol used as a solvent, analytical standard, and research reagent. It has low water solubility, a relatively high boiling point, and is used in partition-coefficient, membrane, and pharmacology studies.
  • Primary alcohol with formula C8H18O and molecular weight 130.23 g/mol.
  • Clear, colorless liquid with low water solubility and a boiling point of about 194-195 °C.
  • Commonly used as a solvent and analytical standard for partition-coefficient and surfactant studies.
  • Reported to inhibit T-type calcium channels in biological research.
  • Supplied in laboratory sizes appropriate for analytical and research use.
Catalog No. 50-002-99357 Supplier Medchemexpress LLC Supplier No. HYW0320131G
1-Octanol (octan-1-ol) is a straight-chain primary fatty alcohol used as a solvent, analytical standard, and research reagent. It has low water solubility, a relatively high boiling point, and is used in partition-coefficient, membrane, and pharmacology studies.
  • Primary alcohol with formula C8H18O and molecular weight 130.23 g/mol.
  • Clear, colorless liquid with low water solubility and a boiling point of about 194-195 °C.
  • Commonly used as a solvent and analytical standard for partition-coefficient and surfactant studies.
  • Reported to inhibit T-type calcium channels in biological research.
  • Supplied in laboratory sizes appropriate for analytical and research use.