Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-octanol | 111-87-5 | MFCD00002988 | 98.0% | 130.23 g/mol | C8H18O | 5 G
SDP

Catalog No. 5000301601
Encompass_Preferred

1-Octanol is a saturated fatty alcohol used as a research reagent and reported T-type calcium channel inhibitor (IC50 ≈4 μM). It is used in pharmacology and analytical applications and is also studied for biofuel properties.

  • Chemical formula: C8H18O; molecular weight: 130.23 g/mol.
  • CAS number: 111-87-5.
  • Purity: approximately 98.0% for research use.
  • Solubility: highly soluble in DMSO (≈100 mg/mL) for in vitro use.
  • Storage: store at room temperature for general stock; in solvent keep at -20 °C to -80 °C for extended periods.

Catalog No. 50-003-01601 Supplier Medchemexpress LLC Supplier No. HYW0320135G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Octanol is a saturated fatty alcohol used as a research reagent and reported T-type calcium channel inhibitor (IC50 ≈4 μM). It is used in pharmacology and analytical applications and is also studied for biofuel properties.

  • Chemical formula: C8H18O; molecular weight: 130.23 g/mol.
  • CAS number: 111-87-5.
  • Purity: approximately 98.0% for research use.
  • Solubility: highly soluble in DMSO (≈100 mg/mL) for in vitro use.
  • Storage: store at room temperature for general stock; in solvent keep at -20 °C to -80 °C for extended periods.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.