Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Octanol (Certified ACS), Fisher Chemical™

Catalog No. a402500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A402-500 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A402-500
This item is not returnable. View return policy

Chemical Identifiers

CAS 111-87-5
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00002988
InChI Key KBPLFHHGFOOTCA-UHFFFAOYSA-N
Synonym 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
PubChem CID 957
ChEBI CHEBI:16188
IUPAC Name octan-1-ol
SMILES CCCCCCCCO

Specifications

Alpha Vector Octanol
Boiling Range 2°C max.
Melting Point -16°C
Color Colorless
Density 0.824g/cm³
Boiling Point 195°C
Flash Point 81°C
Packaging Glass Bottle
Linear Formula CH3(CH2)6CH2OH
Quantity 500 mL
Residue after Evaporation 0.02% max.
Vapor Pressure 0.03mbar at 20°C
Formula Weight 130.23
Grade Certified
Physical Form Liquid
Chemical Name or Material 1-Octanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.