Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Octanol, CHROMASOLV™, for HPLC, ≥99%, Solstice

Catalog No. 6002254 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-022-54 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-022-54 Supplier Solstice Supplier No. 2932451L

missing translation for 'validMainframeMsg'

CHROMASOLV™, for HPLC,≥99%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Specifications

CAS 111-87-5
Absorbance max. 1.0 at 215 nm, max. 0.01 at 400 nm, max. 0.01 at 300 nm, max. 0.05 at 250 nm, max. 0.20 at 235 nm, max. 0.50 at 225 nm
CAS Max % 100.0000%
Melting Point -15°C (literature)
Density 0.827 g/mL at 25 °C(lit.)
Boiling Point 194°C
Flash Point 80°C
Packaging Glass Bottle
Molecular Formula C8H18O
Linear Formula CH3(CH2)7OH
Quantity 1 L
MDL Number MFCD00002988
Beilstein 1697461
Synonym 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
InChI Key KBPLFHHGFOOTCA-UHFFFAOYSA-N
SMILES CCCCCCCCO
IUPAC Name octan-1-ol
Molecular Weight (g/mol) 130.23
PubChem CID 957
ChEBI CHEBI:16188
Formula Weight 130.23
Percent Purity >99%
Grade HPLC
Chemical Name or Material 1-Octanol, CHROMASOLV™, for HPLC, ≥99%
Show More Show Less
ShelfLife 1620 days from date of manufacture
EINECSNumber 203-917-6
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.