Learn More
Medchemexpress LLC 1-octanol-d17 | 153336-13-1 | MFCD00144995 | 99.1% | 147.33 | C8HD17O | 50 MG
SDP
Catalog No. 5000301107
Encompass_Preferred
1-Octanol-d17 is the deuterium-labeled analogue of 1-octanol supplied for research use. It provides deuterium labeling while retaining 1-octanol's chemical properties, enabling use as an isotopic tracer and analytical standard in mass spectrometry, mechanistic studies, and tracer experiments. Characterized by high isotopic and chemical purity, it is intended for laboratory research and analytical applications only.
  • Deuterium-labeled form of 1-octanol for isotopic tracing.
  • High isotopic and chemical purity suitable for mass spectrometry.
  • Useful as an analytical standard and tracer in mechanistic studies.
  • Reported to inhibit T-type calcium channels (native T-currents IC50 ~4 μM).
  • Supplied in small pack sizes appropriate for analytical research.
Catalog No. 50-003-01107 Supplier Medchemexpress LLC Supplier No. HYW032013S50MG
1-Octanol-d17 is the deuterium-labeled analogue of 1-octanol supplied for research use. It provides deuterium labeling while retaining 1-octanol's chemical properties, enabling use as an isotopic tracer and analytical standard in mass spectrometry, mechanistic studies, and tracer experiments. Characterized by high isotopic and chemical purity, it is intended for laboratory research and analytical applications only.
  • Deuterium-labeled form of 1-octanol for isotopic tracing.
  • High isotopic and chemical purity suitable for mass spectrometry.
  • Useful as an analytical standard and tracer in mechanistic studies.
  • Reported to inhibit T-type calcium channels (native T-currents IC50 ~4 μM).
  • Supplied in small pack sizes appropriate for analytical research.