Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-octanol-d17 | 153336-13-1 | MFCD00144995 | 99.1% | 147.33 g/mol | C8HD17O | 5 MG
SDP

Catalog No. 5000300198
Encompass_Preferred

1-Octanol-d17 is the deuterium-labeled analogue of 1-octanol (CAS 153336-13-1) provided as a high-purity isotopically labeled reagent for research and analytical applications. It is commonly used as an internal standard, tracer, or calibration reference in mass spectrometry and analytical chemistry workflows.

  • High isotopic purity for accurate mass spectrometry quantitation.
  • Purity 99.1% to support reliable analytical results.
  • Molecular weight 147.33 g/mol; formula C8HD17O for reference data.
  • Available in small mg pack sizes suitable for analytical workflows.
  • Liquid form with defined density and appearance for straightforward handling.

Catalog No. 50-003-00198 Supplier Medchemexpress LLC Supplier No. HYW032013S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

1-Octanol-d17 is the deuterium-labeled analogue of 1-octanol (CAS 153336-13-1) provided as a high-purity isotopically labeled reagent for research and analytical applications. It is commonly used as an internal standard, tracer, or calibration reference in mass spectrometry and analytical chemistry workflows.

  • High isotopic purity for accurate mass spectrometry quantitation.
  • Purity 99.1% to support reliable analytical results.
  • Molecular weight 147.33 g/mol; formula C8HD17O for reference data.
  • Available in small mg pack sizes suitable for analytical workflows.
  • Liquid form with defined density and appearance for straightforward handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.