Learn More
Medchemexpress LLC 1-octanol-d2 | 78510-02-8 | 99.0% | 132.24 g/mol | C8H16D2O | 25 MG
SDP
Catalog No. 5000302976
Encompass_Preferred
1-Octanol-d2 is the deuterium-labeled analog of 1-octanol provided as a research-grade stable isotope reagent. It is used as an internal standard, tracer, or reference in analytical chemistry and isotopic labeling studies, particularly in mass spectrometry and NMR applications.
  • Deuterated analog suitable as an internal standard for GC-MS and LC-MS analyses.
  • High purity (99.0%), minimizing isotopic impurities.
  • Molecular formula C8H16D2 and molecular weight 132.24 g/mol.
  • Supplied as a small-scale research reagent (25 mg) for analytical workflows.
Catalog No. 50-003-02976 Supplier Medchemexpress LLC Supplier No. 25MGHYW032013S1
1-Octanol-d2 is the deuterium-labeled analog of 1-octanol provided as a research-grade stable isotope reagent. It is used as an internal standard, tracer, or reference in analytical chemistry and isotopic labeling studies, particularly in mass spectrometry and NMR applications.
  • Deuterated analog suitable as an internal standard for GC-MS and LC-MS analyses.
  • High purity (99.0%), minimizing isotopic impurities.
  • Molecular formula C8H16D2 and molecular weight 132.24 g/mol.
  • Supplied as a small-scale research reagent (25 mg) for analytical workflows.