Learn More
Cayman Chemical 1-Octen-3-ol 10g
SDP
Catalog No. 502913722
Encompass_Preferred
A volatile organic compound; an attractant for Ochlerotatus, Psorophora, Anopheles, and Coquillettidia mosquitoes but a repellant for Culex mosquitoes; inhibits dopamine uptake in HEK-DAT cells at 3 and 10 ppm; induces dopaminergic neuron loss and locomotor deficits in, as well as reduces survival of, D. melanogaster at 0.5 ppm via inhalation
Catalog No. 50-291-3722 Supplier Cayman Chemical Supplier No. 3555010 G
A volatile organic compound; an attractant for Ochlerotatus, Psorophora, Anopheles, and Coquillettidia mosquitoes but a repellant for Culex mosquitoes; inhibits dopamine uptake in HEK-DAT cells at 3 and 10 ppm; induces dopaminergic neuron loss and locomotor deficits in, as well as reduces survival of, D. melanogaster at 0.5 ppm via inhalation