Learn More
Medchemexpress LLC 1-Octen-3-ol | 3391-86-4 | 99.97% | 128.22 | 10 G
SDP
Catalog No. 5000451861
Encompass_Preferred
Oct-1-en-3-ol is a fatty acid fragrant and a self-stimulating oxylipin messenger. It acts as a signaling molecule in plant cellular responses, plant-herbivore interactions, and plant-plant interactions. It can also cause dopamine neuron degeneration by disrupting dopamine handling.
  • Fatty acid fragrant.
  • Self-stimulating oxylipin messenger.
  • Signaling molecule in plant cellular responses, plant-herbivore interactions, and plant-plant interactions.
  • Causes dopamine neuron degeneration through disruption of dopamine handling.
  • Initial source: plants, microorganisms.
Catalog No. 50-004-51861 Supplier Medchemexpress LLC Supplier No. HYW01041010G
Oct-1-en-3-ol is a fatty acid fragrant and a self-stimulating oxylipin messenger. It acts as a signaling molecule in plant cellular responses, plant-herbivore interactions, and plant-plant interactions. It can also cause dopamine neuron degeneration by disrupting dopamine handling.
  • Fatty acid fragrant.
  • Self-stimulating oxylipin messenger.
  • Signaling molecule in plant cellular responses, plant-herbivore interactions, and plant-plant interactions.
  • Causes dopamine neuron degeneration through disruption of dopamine handling.
  • Initial source: plants, microorganisms.