Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Octen-3-ol, 98%
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Used as a flavoring agent in cheese and chocolate. Used as a perfuming agent in cosmetic industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as a flavoring agent in cheese and chocolate. Used as a perfuming agent in cosmetic industry.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
Used as a flavoring agent in cheese and chocolate. Used as a perfuming agent in cosmetic industry.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.

Specifications
Specifications
Melting Point | -49°C |
Density | 0.833 |
Boiling Point | 173°C to 175°C |
Flash Point | 61°C (141°F) |
Odor | Characteristic |
Refractive Index | 1.437 |
Quantity | 250 g |
Beilstein | 1744110 |
Solubility Information | Not miscible or difficult to mix in water. |
Formula Weight | 128.22 |
Show More |
Safety and Handling
Hazard Category | H227-H302-H315-H319-H332 |
Hazard Statement | GHS H Statement H302-H315-H319-H335-H227 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
Precautionary Statement | P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P374-P501c |
EINECSNumber | 222-226-0 |
RTECSNumber | RH3300000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction