Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Octen-3-ol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
3391-86-4
C8H16O
128.22
MFCD00004589
VSMOENVRRABVKN-UHFFFAOYSA-N
1-octen-3-ol,vinyl amyl carbinol,1-vinylhexanol,3-hydroxy-1-octene,amyl vinyl carbinol,mushroom alcohol,matsutake alcohol,vinyl hexanol,pentyl vinyl carbinol,matsuica alcohol
18827
CHEBI:34118
oct-1-en-3-ol
CCCCCC(C=C)O
3391-86-4
C8H16O
128.22
MFCD00004589
VSMOENVRRABVKN-UHFFFAOYSA-N
1-octen-3-ol,vinyl amyl carbinol,1-vinylhexanol,3-hydroxy-1-octene,amyl vinyl carbinol,mushroom alcohol,matsutake alcohol,vinyl hexanol,pentyl vinyl carbinol,matsuica alcohol
18827
CHEBI:34118
oct-1-en-3-ol
CCCCCC(C=C)O
Infrared Spectrum Authentic
Boiling Point 174.0°C
Quantity 25g
Density 0.8370g/mL
Color Colorless to Yellow
Flash Point 68°C
Refractive Index 1.4340 to 1.4420
Specific Gravity 0.837
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3(CH2)4CH(OH)CH=CH2
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in ethanol
Formula Weight 128.22
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1-Octen-3-ol
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.