Learn More
Aqua Solutions 1-OCTENE IN SOLVENT ISOPAR E
SDP
Catalog No. NC3404420
Encompass_Preferred
Aqua Solutions 20% w/w 1-octene in Solvent (Isopar E) (600G)
Catalog No. NC3404420 Supplier Aqua Solutions Supplier No. DOW041
Aqua Solutions 20% w/w 1-octene in Solvent (Isopar E) (600G)