Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Octyl-3-methylimidazolium hexafluorophosphate, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
304680-36-2
C12H23N2·PF6
340.29
MFCD03427617
GRCIJNHHTXBJAK-UHFFFAOYSA-N
1-methyl-3-n-octylimidazolium hexafluorophosphate,3-methyl-1-octylimidazolium hexafluorophosphate,1-methyl-3-octylimidazolium hexafluorophosphate,1-octyl-3-methylimidazolium hexafluorophosphate,1-methyl-3-n-octylimidazoliumhexafluorophosphate,acmc-20aj2z,dsstox_cid_27928,dsstox_rid_82681,dsstox_gsid_47952,ksc222c5l
2734243
1-methyl-3-octylimidazol-1-ium;hexafluorophosphate
CCCCCCCCN1C=C[N+](=C1)C.F[P-](F)(F)(F)(F)F
304680-36-2
C12H23N2·PF6
340.29
MFCD03427617
GRCIJNHHTXBJAK-UHFFFAOYSA-N
1-methyl-3-n-octylimidazolium hexafluorophosphate,3-methyl-1-octylimidazolium hexafluorophosphate,1-methyl-3-octylimidazolium hexafluorophosphate,1-octyl-3-methylimidazolium hexafluorophosphate,1-methyl-3-n-octylimidazoliumhexafluorophosphate,acmc-20aj2z,dsstox_cid_27928,dsstox_rid_82681,dsstox_gsid_47952,ksc222c5l
2734243
1-methyl-3-octylimidazol-1-ium;hexafluorophosphate
CCCCCCCCN1C=C[N+](=C1)C.F[P-](F)(F)(F)(F)F
Ionic liquid
Viscosity 866 mPa.s (20°C)
Specific Gravity 1.2345
Refractive Index 1.4230 to 1.4250
Packaging Glass bottle
Quantity 5g
Density 1.2345g/mL
Color Yellow
NMR 98.0% min.
Infrared Spectrum Authentic
Water 1% Max. (K.F.)
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Formula Weight 340.29
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material 1-Octyl-3-methylimidazolium hexafluorophosphate
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA