Learn More
Medchemexpress LLC 1-Oleoyl-2-hydroxy-sn-glycero-3-pg (sodium) | 326495-24-3 | 99.0% | C24H46NaO9P | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1455051MG
Encompass_Preferred
1-Oleoyl-2-hydroxy-sn-glycero-3-PG sodium is a lysophospholipid characterized by an oleic acid (18:1) at the sn-1 position. This product is suitable for generating micelles, liposomes, and various artificial membranes, including lipid-based drug carrier systems, and is intended for research use only.
  • Contains oleic acid (18:1) at the sn-1 position.
  • Used in the generation of micelles and liposomes.
  • Suitable for lipid-based drug carrier systems.
  • For research use only.
Catalog No. 50-003-53809