Learn More
Medchemexpress LLC 1-oleoyl-2-hydroxy-sn-glycero-3-phosphoglycerol sodium | 326495-24-3 | 99.0% | C24H46NaO9P | 10MG

Supplier: Medchemexpress LLC HY14550510MG
1-Oleoyl-2-hydroxy-sn-glycero-3-phosphoglycerol sodium is a lysophospholipid reagent containing an oleoyl (18:1) chain at the sn-1 position. Supplied as a solid sodium salt for research use, it is characterized by high purity and is used in studies of lipid signaling, membrane biology, and biochemical assays.
- Lysophospholipid with oleoyl (18:1) at sn-1
- Sodium salt form, solid
- High purity (99.0%)
- Molecular weight 532.58 g·mol⁻¹
- Storage: store sealed at -20°C; in solvent -80°C up to 6 months
- Suitable for lipid signaling and membrane biology studies
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.