Learn More
Medchemexpress LLC 1-oleoyl-2-palmitoyl-sn-glycero-3-phosphocholine | 59491-62-2 | MFCD00674332 | 99.8% | 760.08 | C42H82NO8P | 5 MG
SDP
Catalog No. 5000220987
Encompass_Preferred
1-Oleoyl-2-palmitoyl-sn-glycero-3-phosphocholine is a zwitterionic phospholipid used as a lipid component in model membranes and for preparing liposomes and other lipid assemblies in biochemical and biophysical research.
  • High purity suitable for research applications (99.8%).
  • White to off-white solid appearance and consistent quality.
  • Molecular formula C42H82NO8P; molecular weight 760.08 g/mol.
  • Suitable for liposome synthesis and membrane model systems.
  • Powder storage: -20°C for up to 3 years; in solvent: -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-002-20987 Supplier Medchemexpress LLC Supplier No. HY1429905MG
1-Oleoyl-2-palmitoyl-sn-glycero-3-phosphocholine is a zwitterionic phospholipid used as a lipid component in model membranes and for preparing liposomes and other lipid assemblies in biochemical and biophysical research.
  • High purity suitable for research applications (99.8%).
  • White to off-white solid appearance and consistent quality.
  • Molecular formula C42H82NO8P; molecular weight 760.08 g/mol.
  • Suitable for liposome synthesis and membrane model systems.
  • Powder storage: -20°C for up to 3 years; in solvent: -80°C for 6 months, -20°C for 1 month.