Learn More
Medchemexpress LLC 1-oleoyl-2-palmitoyl-sn-glycero-3-phosphocholine | 59491-62-2 | MFCD00674332 | 99.8% | C42H82NO8P | 10MG
SDP
Catalog No. 5000261128
Encompass_Preferred
1-Oleoyl-2-palmitoyl-sn-glycero-3-phosphocholine is a phosphatidylcholine lipid used to prepare liposomes and artificial membranes for membrane biophysics, toxin interaction studies, and enzymology. It is a white to off-white solid (C42H82NO8P, MW 760.1 g/mol), typically supplied at high purity and recommended to be stored at -20°C for stability.
  • High purity suitable for biochemical applications.
  • Ideal for liposome and artificial membrane preparation.
  • Solid form allows easy weighing and formulation.
  • Compatible with studies of membrane proteins, toxins, and phospholipases.
  • Stable under recommended frozen storage.
Catalog No. 50-002-61128 Supplier Medchemexpress LLC Supplier No. HY14299010MG
1-Oleoyl-2-palmitoyl-sn-glycero-3-phosphocholine is a phosphatidylcholine lipid used to prepare liposomes and artificial membranes for membrane biophysics, toxin interaction studies, and enzymology. It is a white to off-white solid (C42H82NO8P, MW 760.1 g/mol), typically supplied at high purity and recommended to be stored at -20°C for stability.
  • High purity suitable for biochemical applications.
  • Ideal for liposome and artificial membrane preparation.
  • Solid form allows easy weighing and formulation.
  • Compatible with studies of membrane proteins, toxins, and phospholipases.
  • Stable under recommended frozen storage.