Learn More
Cayman Chemical 1-Oleoyl Glycerol, Purity: ≥95%, CAS: 111-03-5, 1 g
SDP
Catalog No. 502932947
Encompass_Preferred
A monoacylglycerol; enhances diffusion of cyclosporin A from the donor compartment of a Franz diffusion cell into the stratum corneum, epidermis, and dermis of porcine ear skin, as well as through the skin to the receptor compartment when used in a formulation with cyclosporin A; Mixed micellar systems containing 1-oleoyl glycerol and bile salts increase Papp and absorption rate of amphotericine B in perfused rat intestine in vivo; LNPs containing 1-oleoyl-glycerol and encapsulating various mRNAs exhibit enhanced pulmonary delivery via intranasal administration and targeted spleen delivery via intravenous administration in mice compared with LNPs containing DSPC
Catalog No. 50-293-2947 Supplier Cayman Chemical Supplier No. 267931 G
A monoacylglycerol; enhances diffusion of cyclosporin A from the donor compartment of a Franz diffusion cell into the stratum corneum, epidermis, and dermis of porcine ear skin, as well as through the skin to the receptor compartment when used in a formulation with cyclosporin A; Mixed micellar systems containing 1-oleoyl glycerol and bile salts increase Papp and absorption rate of amphotericine B in perfused rat intestine in vivo; LNPs containing 1-oleoyl-glycerol and encapsulating various mRNAs exhibit enhanced pulmonary delivery via intranasal administration and targeted spleen delivery via intravenous administration in mice compared with LNPs containing DSPC