Learn More
Medchemexpress LLC 1-oleoyl lysophosphatidic acid (1-oleoyl-sn-glycero-3-phosphate) sodium | 325465-93-8 | MFCD00133427 | 99.8% | 458.50 | C21H40NaO7P | 100 MG
SDP
Catalog No. 5000241616
Encompass_Preferred
1-Oleoyl lysophosphatidic acid sodium is the sodium salt of a bioactive lysophospholipid that functions as an agonist at lysophosphatidic acid (LPA) receptors. It is supplied as a solid research reagent for cell signaling and biochemical assays, commonly used to study LPA-mediated pathways and receptor activation.
  • Acts as an agonist of LPA receptors for receptor activation studies.
  • Water-soluble sodium salt for improved handling and formulation.
  • High purity suitable for in vitro signaling and biochemical assays.
  • Available in multiple package sizes for small- to moderate-scale experiments.
  • Stable when stored frozen to preserve activity.
Catalog No. 50-002-41616 Supplier Medchemexpress LLC Supplier No. HY107614100MG
1-Oleoyl lysophosphatidic acid sodium is the sodium salt of a bioactive lysophospholipid that functions as an agonist at lysophosphatidic acid (LPA) receptors. It is supplied as a solid research reagent for cell signaling and biochemical assays, commonly used to study LPA-mediated pathways and receptor activation.
  • Acts as an agonist of LPA receptors for receptor activation studies.
  • Water-soluble sodium salt for improved handling and formulation.
  • High purity suitable for in vitro signaling and biochemical assays.
  • Available in multiple package sizes for small- to moderate-scale experiments.
  • Stable when stored frozen to preserve activity.