Learn More
Medchemexpress LLC 1-oleoyl lysophosphatidic acid (sodium) | 325465-93-8 | MFCD00133427 | 99.8% | 458.50 | C21H40NaO7P | 5 MG
SDP
Catalog No. 5000240437
Encompass_Preferred
1-Oleoyl lysophosphatidic acid sodium is a bioactive lysophospholipid that functions as a potent agonist of lysophosphatidic acid (LPA) receptors. It is supplied as a high-purity research reagent for studies of GPCR signaling, cell proliferation, migration, and neural differentiation. The sodium salt form improves aqueous handling for in vitro and in vivo formulations.
Catalog No. 50-002-40437 Supplier Medchemexpress LLC Supplier No. HY1076145MG
1-Oleoyl lysophosphatidic acid sodium is a bioactive lysophospholipid that functions as a potent agonist of lysophosphatidic acid (LPA) receptors. It is supplied as a high-purity research reagent for studies of GPCR signaling, cell proliferation, migration, and neural differentiation. The sodium salt form improves aqueous handling for in vitro and in vivo formulations.