Learn More
Medchemexpress LLC 1-oleoyl lysophosphatidic acid sodium | 325465-93-8 | MFCD00133427 | 99.8% | 458.50 g/mol | C21H40NaO7P | 50 MG
SDP
Catalog No. 5000240388
Encompass_Preferred
1-Oleoyl lysophosphatidic acid sodium is a bioactive lysophospholipid research reagent that activates lysophosphatidic acid (LPA) receptors and is used in cell-based studies of signaling, proliferation, and neurological responses.
  • Acts as a potent agonist of LPA receptors.
  • Promotes DNA synthesis and cell proliferation.
  • High purity (99.8%).
  • Chemical formula C21H40NaO7P; molecular weight 458.50 g/mol.
  • Solid form; store at -20°C, in solvent store at -80°C for long-term.
  • Available in small research pack sizes, including 50 MG.
Catalog No. 50-002-40388 Supplier Medchemexpress LLC Supplier No. HY10761450MG
1-Oleoyl lysophosphatidic acid sodium is a bioactive lysophospholipid research reagent that activates lysophosphatidic acid (LPA) receptors and is used in cell-based studies of signaling, proliferation, and neurological responses.
  • Acts as a potent agonist of LPA receptors.
  • Promotes DNA synthesis and cell proliferation.
  • High purity (99.8%).
  • Chemical formula C21H40NaO7P; molecular weight 458.50 g/mol.
  • Solid form; store at -20°C, in solvent store at -80°C for long-term.
  • Available in small research pack sizes, including 50 MG.