Learn More
Cayman Chemical 1-Oleoyl Lysophosphatidic Acid (sodium salt), 22556-62-3, 10 mg
SDP
Supplier:  Cayman Chemical 6221510
Encompass_Preferred
1-Oleoyl lysophosphatidic acid is a species of LPA containing oleic acid at the sn-1 position. Phosphatidic acid is produced either directly through the action of PLD or through a two step process involving liberation of DAG by PLC followed by phosphorylation of DAG by diglycerol kinase.{2656} Hydrolysis of the fatty acid at the sn-2 position by PLA2 yields bioactive LPA. LPA binds to one of four different G-protein linked receptors{13001} to mediate a variety of biological responses including cell proliferation, smooth muscle contraction, platelet aggregation, neurite retraction, and cell motility.{2656} 1-Oleoyl lysophosphatidic acid is the most potent of the LPA analogs for calcium mobilization in A431 cells{2665} and for growth stimulation of a variety of cell lines.{2666}
Catalog No. NC9491394
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.