Learn More
Medchemexpress LLC 1-oleoyl-sn-glycero-3-phosphocholine | 19420-56-5 | MFCD00133435 | 99.6% | 521.67 | C26H52NO7P | 1 MG
SDP
Catalog No. 5000282856
Encompass_Preferred
1-Oleoyl-sn-glycero-3-phosphocholine is an unsaturated lysophosphatidylcholine used as an analytical standard and reagent in lipid research, biochemical assays, and method development. Supplied as a white to off-white solid with high purity, it is intended for analytical and research use and requires low-temperature storage for long-term stability.
  • High purity analytical standard (99.6%).
  • Available as small solid quantities and as ready-to-use DMSO solution.
  • Suitable for lipid analysis, assay development, and reference standard applications.
  • White to off-white solid that is soluble in common organic solvents such as DMSO.
  • Stable as a powder at -20°C; limited stability in solution, follow storage guidance.
Catalog No. 50-002-82856 Supplier Medchemexpress LLC Supplier No. HYW1404391MG
1-Oleoyl-sn-glycero-3-phosphocholine is an unsaturated lysophosphatidylcholine used as an analytical standard and reagent in lipid research, biochemical assays, and method development. Supplied as a white to off-white solid with high purity, it is intended for analytical and research use and requires low-temperature storage for long-term stability.
  • High purity analytical standard (99.6%).
  • Available as small solid quantities and as ready-to-use DMSO solution.
  • Suitable for lipid analysis, assay development, and reference standard applications.
  • White to off-white solid that is soluble in common organic solvents such as DMSO.
  • Stable as a powder at -20°C; limited stability in solution, follow storage guidance.