Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-oleoyl-sn-glycero-3-phosphocholine | 19420-56-5 | MFCD00133435 | 99.6% | 521.67 g/mol | C26H52NO7P | 100 MG
SDP

Catalog No. 5000283461
Encompass_Preferred

1-Oleoyl-sn-glycero-3-phosphocholine is an unsaturated lysophosphatidylcholine (18:1 Lyso PC) used as a biochemical reagent and analytical standard in lipid research and lipidomics. It is supplied as a high-purity solid suitable for analytical workflows and method development.

  • High purity (99.6%) for analytical use.
  • White to off-white solid appearance, easy to handle.
  • Suitable as a lipidomics standard and biochemical reagent.
  • Molecular formula C26H52NO7P and molecular weight 521.67 g/mol.
  • CAS number 19420-56-5 for unambiguous identification.
  • Available in common laboratory quantities including 100 MG.

Catalog No. 50-002-83461 Supplier Medchemexpress LLC Supplier No. HYW140439100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Oleoyl-sn-glycero-3-phosphocholine is an unsaturated lysophosphatidylcholine (18:1 Lyso PC) used as a biochemical reagent and analytical standard in lipid research and lipidomics. It is supplied as a high-purity solid suitable for analytical workflows and method development.

  • High purity (99.6%) for analytical use.
  • White to off-white solid appearance, easy to handle.
  • Suitable as a lipidomics standard and biochemical reagent.
  • Molecular formula C26H52NO7P and molecular weight 521.67 g/mol.
  • CAS number 19420-56-5 for unambiguous identification.
  • Available in common laboratory quantities including 100 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.