Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Oxa-2-oxo-3-thiaindolizinium Chloride 98.0+%, TCI America™
SDP

Catalog No. O02705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02705G 5 g
O02701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02705G Supplier TCI America Supplier No. O02705G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 89025-51-4 | C6H5ClNO2S+ | 190.621 g/mol

Chemical Identifiers

CAS 89025-51-4
Molecular Formula C6H5ClNO2S+
Molecular Weight (g/mol) 190.621
MDL Number MFCD00134271
InChI Key PVTYXZXONFHZCV-UHFFFAOYSA-N
Synonym 2-Oxo[1.4.2]oxathiazolo[2,3-a]pyridinium Chloride
PubChem CID 91873692
IUPAC Name [1,3,4]oxathiazolo[4,5-a]pyridin-4-ium-2-one;hydrochloride
SMILES C1=CC=[N+]2C(=C1)OC(=O)S2.Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 189.61
Percent Purity ≥98.0% (N)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material 1-Oxa-2-oxo-3-thiaindolizinium Chloride [for Source of Alkyl Radical]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.