Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Oxa-6-azaspiro[3.4]octane oxalate, 96%

Catalog No. AAH57452ME
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57452ME 500 mg
AAH57452MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH57452ME Supplier Thermo Scientific Chemicals Supplier No. H57452ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1408074-51-0 | C8H13NO5 | 203.19 g/mol

1-Oxa-6-azaspiro[3.4]octane oxalate is used as a pharmaceutical intermediate in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Oxa-6-azaspiro[3.4]octane oxalate is used as a pharmaceutical intermediate in synthetic chemistry.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1408074-51-0
Molecular Formula C8H13NO5
Molecular Weight (g/mol) 203.19
MDL Number MFCD19982774
InChI Key ZKTJFOHJBNDMDS-UHFFFAOYNA-N
Synonym 1-oxa-6-azaspiro 3.4 octane oxalate, 1-oxa-6-azaspiro 3.4 octane; oxalic acid
PubChem CID 71432930
IUPAC Name 1-oxa-7-azaspiro[3.4]octane;oxalic acid
SMILES OC(=O)C(O)=O.C1CC2(CCNC2)O1

Specifications

Quantity 500 mg
Formula Weight 203.19
Percent Purity 96%
Chemical Name or Material 1-Oxa-6-azaspiro[3.4]octane oxalate
Hazard Category H302+H312
Hazard Statement GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.