Learn More
Cayman Chemical 1-oxo Ibuprofen, Purity: ≥98%, CAS: 65813-55-0, 10 mg
SDP
Catalog No. 502986774
Encompass_Preferred
A degradation product and potential impurity in commercial ibuprofen preparations
Catalog No. 50-298-6774 Supplier Cayman Chemical Supplier No. 2257310 MG
A degradation product and potential impurity in commercial ibuprofen preparations